Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zdhhc7
Entrez Gene ID
436875
Gene Name
zinc finger DHHC-type palmitoyltransferase 7
Gene Aliases
zgc:92305
Chromosome Location
Chr.18: 5813996 - 5830417 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002602.1 NP_001002602.1 3 - 4 526 60
GenBank mRNA BC075993.1 - 3 - 4 526 60

View Full Product Details