Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zdhhc7
Entrez Gene ID
436875
Gene Name
zinc finger DHHC-type palmitoyltransferase 7
Gene Aliases
zgc:92305
Chromosome Location
Chr.18: 5813996 - 5830417 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002602.1 NP_001002602.1 4 - 5 650 119
GenBank mRNA BC075993.1 - 4 - 5 650 119

View Full Product Details