Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bcl7bb
Entrez Gene ID
81595
Gene Name
BAF chromatin remodeling complex subunit BCL7B b
Gene Aliases
bcl7b, zgc:103501
Chromosome Location
Chr.18: 62399 - 72052 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006018.1 NP_001006018.1 4 - 5 509 68
GenBank mRNA BC083405.1 - 4 - 5 509 68
BC165913.1 - 4 - 5 489 68

View Full Product Details