Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gsna
Entrez Gene ID
321099
Gene Name
gelsolin a
Gene Aliases
cb107, gsn, sb:cb107, u-gelsolin, wu:fi16f06
Chromosome Location
Chr.21: 17569613 - 17603199 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012312.2 NP_001012312.2 11 - 12 1686 89
XM_021468752.1 XP_021324427.1 12 - 13 1737 89
GenBank mRNA AY731180.1 - 11 - 12 1702 89
BC125892.1 - 11 - 12 1665 89
CN326697.1 - 4 - 5 435 89

View Full Product Details