Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fn1b
Entrez Gene ID
334613
Gene Name
fibronectin 1b
Gene Aliases
cb1057, fn1l, fn3, wu:fa14f11, wu:fb03c02
Chromosome Location
Chr.1: 5504131 - 5543086 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013261.1 NP_001013279.1 41 - 42 6894 77
XM_005160284.3 XP_005160341.1 41 - 42 6926 77
XM_009305554.3 XP_009303829.1 42 - 43 7206 77
GenBank mRNA AF500198.1 - 3 - 4 671 77
AY193829.1 - 3 - 4 422 77
AY538257.1 - 41 - 42 6894 77
AY725818.1 - 42 - 43 7185 77

View Full Product Details