Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hgfa
Entrez Gene ID
493781
Gene Name
hepatocyte growth factor a
Gene Aliases
hgf1, si:dkey-146j3.2
Chromosome Location
Chr.4: 19710681 - 19743229 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013274.1 NP_001013292.1 3 - 4 475 62
XM_005164742.3 XP_005164799.1 3 - 4 1401 62
GenBank mRNA BC135005.1 - 3 - 4 478 62

View Full Product Details