Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccnh
Entrez Gene ID
541325
Gene Name
cyclin H
Gene Aliases
zgc:114132
Chromosome Location
Chr.5: 47963938 - 47975440 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013453.1 NP_001013471.1 3 - 4 420 116
XM_005165512.4 XP_005165569.1 3 - 4 408 116
XM_021476140.1 XP_021331815.1 3 - 4 489 116
GenBank mRNA BC091559.1 - 3 - 4 420 116
DQ294346.1 - 3 - 4 400 116

View Full Product Details