Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccnh
Entrez Gene ID
541325
Gene Name
cyclin H
Gene Aliases
zgc:114132
Chromosome Location
Chr.5: 47963938 - 47975440 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013453.1 NP_001013471.1 8 - 9 1041 110
XM_005165512.4 XP_005165569.1 8 - 9 1029 110
XM_021476140.1 XP_021331815.1 8 - 9 1110 110
GenBank mRNA BC091559.1 - 8 - 9 1041 110
DQ294346.1 - 8 - 9 1021 110

View Full Product Details