Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gmpr
Entrez Gene ID
553639
Gene Name
guanosine monophosphate reductase
Gene Aliases
zgc:110617
Chromosome Location
Chr.19: 227860 - 230081 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020612.1 NP_001018448.1 2 - 3 277 67
GenBank mRNA BC095337.1 - 2 - 3 277 67
BC164152.1 - 2 - 3 242 67

View Full Product Details