Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gmpr
Entrez Gene ID
553639
Gene Name
guanosine monophosphate reductase
Gene Aliases
zgc:110617
Chromosome Location
Chr.19: 227860 - 230081 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020612.1 NP_001018448.1 6 - 7 723 82
GenBank mRNA BC095337.1 - 6 - 7 723 82
BC164152.1 - 6 - 7 688 82

View Full Product Details