Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgrb
Entrez Gene ID
554142
Gene Name
retinal G protein coupled receptor b
Gene Aliases
rgr2, wu:fb64c10, zgc:109930, zgc:92677
Chromosome Location
Chr.12: 48965034 - 48970458 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024436.1 NP_001019607.1 4 - 4 672 63
GenBank mRNA BC076161.1 - 4 - 4 559 63
BC095058.1 - 4 - 4 672 63
KT008427.1 - 4 - 4 513 63

View Full Product Details