Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rhd
Entrez Gene ID
554272
Gene Name
Rh blood group, D antigen
Gene Aliases
rh30
Chromosome Location
Chr.13: 45496766 - 45523026 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024819.1 NP_001019990.1 8 - 9 1207 68
GenBank mRNA AY613958.1 - 8 - 9 1207 68

View Full Product Details