Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp2r2ca
Entrez Gene ID
100002802
Gene Name
protein phosphatase 2, regulatory subunit B, gamma a
Gene Aliases
ppp2r2c, zgc:123166
Chromosome Location
Chr.1: 14283692 - 14306853 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037394.1 NP_001032471.1 9 - 10 1357 92
XM_021478641.1 XP_021334316.1 8 - 9 1416 92
GenBank mRNA BC109406.1 - 9 - 10 1357 92

View Full Product Details