Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- tnikb
- Entrez Gene ID
-
556959
- Gene Name
- TRAF2 and NCK interacting kinase b
- Gene Aliases
-
im:6898857, zgc:123033, zgc:152783
- Chromosome Location
-
Chr.24: 26488367 - 26632227 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC108020.1 |
- |
11 - 12 |
1501 |
144 |
View Full Product Details