Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fstb
Entrez Gene ID
566538
Gene Name
follistatin b
Gene Aliases
fst2, fstl, hm:zeh1606, zeh1606, zgc:171342
Chromosome Location
Chr.10: 35059 - 43676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039631.1 NP_001034720.1 5 - 6 1020 66
GenBank mRNA DQ317969.1 - 5 - 6 1020 66

View Full Product Details