Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
olfml2ba
Entrez Gene ID
557997
Gene Name
olfactomedin-like 2Ba
Gene Aliases
si:ch211-199m3.8
Chromosome Location
Chr.20: 33791944 - 33810984 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044849.1 NP_001038314.1 1 - 2 151 82
XM_005160712.4 XP_005160769.1 1 - 2 363 82
XM_009294746.3 XP_009293021.1 1 - 2 363 82

View Full Product Details