Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alx1
Entrez Gene ID
565176
Gene Name
ALX homeobox 1
Gene Aliases
CART-1, cart1, si:ch211-154h20.1
Chromosome Location
Chr.18: 156730 - 164673 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045074.1 NP_001038539.1 1 - 2 205 91

View Full Product Details