Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fcer1g
Entrez Gene ID
100101464
Gene Name
Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide
Gene Aliases
FcRgamma
Chromosome Location
Chr.7: 19033883 - 19043391 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100106.2 NP_001093576.2 3 - 4 243 118
GenBank mRNA EF158447.1 - 3 - 4 256 118

View Full Product Details