Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vtg2
Entrez Gene ID
559931
Gene Name
vitellogenin 2
Gene Aliases
si:dkey-4c23.2, vg2, zfVtg2
Chromosome Location
Chr.22: 24869690 - 24880824 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110384.1 NP_001103854.1 13 - 14 1410 131
GenBank mRNA BC154732.1 - 13 - 14 1410 131

View Full Product Details