Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pth1r
Entrez Gene ID
30629
Gene Name
parathyroid hormone 1 receptor
Gene Aliases
pth1ra, pthr1, zPTH1R
Chromosome Location
Chr.2: 2290148 - 2451181 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131357.1 NP_571432.1 2 - 3 322 68
GenBank mRNA AF132084.1 - 2 - 3 322 68
BC162211.1 - 2 - 3 68
BC162544.1 - 2 - 3 68

View Full Product Details