Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naca
Entrez Gene ID
195823
Gene Name
nascent polypeptide associated complex subunit alpha
Gene Aliases
ik:tdsubc_2b5, nkn, tdsubc_2b5, unm_t30953, xx:tdsubc_2b5
Chromosome Location
Chr.23: 17866008 - 17874341 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193352.1 NP_001180281.1 6 - 7 2936 83
NM_173264.2 NP_775371.1 5 - 6 491 83
GenBank mRNA AY099510.1 - 5 - 6 427 83
AY398384.1 - 5 - 6 431 83
BC122432.1 - 5 - 6 476 83
BC164312.1 - 4 - 5 419 83

View Full Product Details