Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zmynd19
Entrez Gene ID
325735
Gene Name
zinc finger, MYND-type containing 19
Gene Aliases
MIZIP, fd07e10, wu:fd07e10
Chromosome Location
Chr.5: 54337284 - 54395397 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183069.1 NP_898892.1 4 - 5 501 66
GenBank mRNA BC045296.1 - 4 - 5 501 66
BC165425.1 - 4 - 5 390 66

View Full Product Details