Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pif1
Entrez Gene ID
326729
Gene Name
PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae)
Gene Aliases
cb934, fe11d03, wu:fe11d03, zgc:56161
Chromosome Location
Chr.2: 17096850 - 17114853 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198807.1 NP_942102.1 11 - 12 1744 60
GenBank mRNA BC045956.1 - 11 - 12 1744 60

View Full Product Details