Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uck2b
Entrez Gene ID
327057
Gene Name
uridine-cytidine kinase 2b
Gene Aliases
wu:fa20g04, zgc:56174
Chromosome Location
Chr.2: 6051916 - 6065416 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199764.2 NP_956058.1 2 - 3 363 65
GenBank mRNA BC045968.1 - 2 - 3 308 65
BC164836.1 - 2 - 3 299 65
CB357915.1 - 2 - 3 298 65

View Full Product Details