Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
maco1b
Entrez Gene ID
393435
Gene Name
macoilin 1b
Gene Aliases
tmem57b, zgc:63610
Chromosome Location
Chr.13: 45524388 - 45570504 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200463.2 NP_956757.2 3 - 4 471 140
XM_005157048.4 XP_005157105.1 3 - 4 450 140
XM_021480543.1 XP_021336218.1 3 - 4 558 140
GenBank mRNA AY845030.1 - 3 - 4 471 140
BC055164.1 - 4 - 5 419 140
BC171392.1 - 3 - 4 450 140

View Full Product Details