Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
agfg1b
Entrez Gene ID
393560
Gene Name
ArfGAP with FG repeats 1b
Gene Aliases
zgc:66055
Chromosome Location
Chr.2: 16460305 - 16479291 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200588.2 NP_956882.2 2 - 3 327 84
XM_005163316.4 XP_005163373.1 2 - 3 422 84
GenBank mRNA BC056733.1 - 2 - 3 327 84

View Full Product Details