Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
calb2a
Entrez Gene ID
393691
Gene Name
calbindin 2a
Gene Aliases
calb2l, wu:fq18e08, zgc:73115
Chromosome Location
Chr.18: 177730 - 198733 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200718.1 NP_957012.1 9 - 10 683 82
GenBank mRNA BC059479.1 - 9 - 10 683 82

View Full Product Details