Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naprt
Entrez Gene ID
393864
Gene Name
nicotinate phosphoribosyltransferase
Gene Aliases
naprt1, zgc:77870
Chromosome Location
Chr.20: 52273446 - 52314347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200890.2 NP_957184.2 10 - 11 1435 69
XM_005170215.4 XP_005170272.1 10 - 11 1442 69
GenBank mRNA BC063987.1 - 10 - 11 1397 69
EH475595.1 - 3 - 4 617 69

View Full Product Details