Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
naprt
Entrez Gene ID
393864
Gene Name
nicotinate phosphoribosyltransferase
Gene Aliases
naprt1, zgc:77870
Chromosome Location
Chr.20: 52273446 - 52314347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200890.2 NP_957184.2 11 - 12 1595 59
XM_005170215.4 XP_005170272.1 11 - 12 1602 59
GenBank mRNA BC063987.1 - 11 - 12 1557 59
EH475595.1 - 2 - 3 457 59

View Full Product Details