Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sigmar1
Entrez Gene ID
393952
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
oprs1, zgc:56378
Chromosome Location
Chr.10: 14488625 - 14494245 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200977.1 NP_957271.1 2 - 3 443 98
GenBank mRNA BC049416.1 - 2 - 3 443 98

View Full Product Details