Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
naa15b
Entrez Gene ID
334099
Gene Name
N-alpha-acetyltransferase 15, NatA auxiliary subunit b
Gene Aliases
fi31d06, narg1b, wu:fi31d06
Chromosome Location
Chr.1: 40613297 - 40641474 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203321.2 NP_976066.2 1 - 2 222 99
GenBank mRNA AY391464.1 - 1 - 2 209 99
BC044392.1 - 1 - 2 139 99
BC071438.1 - 1 - 2 222 99

View Full Product Details