Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
relt
Entrez Gene ID
404605
Gene Name
RELT TNF receptor
Gene Aliases
Tnfrsf19l, zgc:77246
Chromosome Location
Chr.18: 2603628 - 2668659 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207073.1 NP_996956.1 2 - 3 187 100
GenBank mRNA BC066389.1 - 2 - 3 187 100

View Full Product Details