Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rbbp4
Entrez Gene ID
321726
Gene Name
retinoblastoma binding protein 4
Gene Aliases
rbb4, rbb4-2, wu:fb33a09, wu:fb40e10, zgc:55349, zgc:77854
Chromosome Location
Chr.19: 30999839 - 31007318 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212595.1 NP_997760.1 8 - 9 1123 126
GenBank mRNA AY394940.1 - 8 - 9 1123 126
BC045315.1 - 8 - 9 1057 126
BC063984.1 - 8 - 9 1036 126
BC155193.1 - 8 - 9 1071 126
BC165845.1 - 8 - 9 996 126

View Full Product Details