Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
actr1
Entrez Gene ID
406681
Gene Name
actin related protein 1, centractin
Gene Aliases
Actr1b, hm:zehn1110, wu:fc22g09, wu:fc56d01, wu:fy52e08, zehn1110, zgc:56317
Chromosome Location
Chr.10: 42521970 - 42540839 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213372.1 NP_998537.1 7 - 8 896 123
GenBank mRNA BC046069.1 - 7 - 8 896 123
BC097070.1 - 7 - 8 887 123

View Full Product Details