Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpf2
Entrez Gene ID
406587
Gene Name
ribosome production factor 2 homolog
Gene Aliases
bxdc1, ik:tdsubc_1a3, wu:fe17d08, wu:fj16d06, xx:tdsubc_1a3, zgc:77150
Chromosome Location
Chr.20: 150921 - 154990 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214748.1 NP_999913.1 5 - 6 355 76
GenBank mRNA BC065672.1 - 5 - 6 355 76

View Full Product Details