Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fntb
Entrez Gene ID
323500
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
wu:fb99h09, zgc:86774
Chromosome Location
Chr.20: 28803961 - 28827769 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002128.1 NP_001002128.1 10 - 11 1091 66
XM_021468366.1 XP_021324041.1 9 - 10 1028 66
GenBank mRNA BC071443.1 - 10 - 11 1091 66

View Full Product Details