Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fntb
Entrez Gene ID
323500
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
wu:fb99h09, zgc:86774
Chromosome Location
Chr.20: 28803961 - 28827769 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002128.1 NP_001002128.1 5 - 6 548 66
XM_021468366.1 XP_021324041.1 5 - 6 572 66
GenBank mRNA BC071443.1 - 5 - 6 548 66

View Full Product Details