Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dctn5
Entrez Gene ID
436770
Gene Name
dynactin 5
Gene Aliases
zgc:92842
Chromosome Location
Chr.3: 35550870 - 35554801 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002497.1 NP_001002497.1 5 - 6 523 143
GenBank mRNA BC076304.1 - 5 - 6 523 143
BC164967.1 - 5 - 6 486 143

View Full Product Details