Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnaseh1
Entrez Gene ID
436932
Gene Name
ribonuclease H1
Gene Aliases
zgc:91971
Chromosome Location
Chr.20: 30378858 - 30385570 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002659.2 NP_001002659.2 4 - 5 713 69
NM_001285828.1 NP_001272757.1 5 - 6 531 69
GenBank mRNA BC076457.1 - 4 - 5 694 69
BC164625.1 - 4 - 5 509 69
EH544014.1 - 5 - 6 554 69

View Full Product Details