Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnaseh1
Entrez Gene ID
436932
Gene Name
ribonuclease H1
Gene Aliases
zgc:91971
Chromosome Location
Chr.20: 30378858 - 30385570 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002659.2 NP_001002659.2 6 - 7 858 88
NM_001285828.1 NP_001272757.1 7 - 8 676 88
GenBank mRNA BC076457.1 - 6 - 7 839 88
BC164625.1 - 6 - 7 654 88
EH544014.1 - 7 - 8 699 88

View Full Product Details