Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arrdc3b
Entrez Gene ID
447866
Gene Name
arrestin domain containing 3b
Gene Aliases
zgc:92034
Chromosome Location
Chr.10: 43761480 - 43771447 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004605.1 NP_001004605.1 1 - 2 434 110
GenBank mRNA BC078434.1 - 1 - 2 434 110

View Full Product Details