Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kif7
Entrez Gene ID
544651
Gene Name
kinesin family member 7
Gene Aliases
cos2, costal2
Chromosome Location
Chr.7: 14394515 - 14446588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014816.1 NP_001014816.1 8 - 9 2066 90
XM_005166379.4 XP_005166436.1 9 - 10 2237 90
GenBank mRNA AY954727.1 - 8 - 9 2066 90

View Full Product Details