Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rtn1a
Entrez Gene ID
323706
Gene Name
reticulon 1a
Gene Aliases
fb64c01, im:7136397, rtn1, rtn5, wu:fb64c01, wu:fc07e07, wu:fq24d12, zgc:113566, zgc:73298, zgc:86778
Chromosome Location
Chr.13: 31424586 - 31470469 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029967.3 NP_001025138.2 5 - 6 2461 80
NM_001319289.1 NP_001306218.1 3 - 4 447 80
NM_199652.2 NP_955946.1 3 - 4 700 80
GenBank mRNA AY555044.1 - 5 - 6 2364 80
AY555046.1 - 5 - 6 926 80
AY555049.1 - 3 - 4 539 80
AY555050.1 - 3 - 4 576 80
BC059623.1 - 3 - 4 554 80
BC071446.1 - 3 - 4 497 80
BC090486.1 - 3 - 4 992 80
BC163936.1 - 5 - 6 2431 80
EH444584.1 - 3 - 4 650 80
EH487664.1 - 3 - 4 412 80
EH494741.1 - 3 - 4 473 80
EH594768.1 - 3 - 4 667 80

View Full Product Details