Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgi3
Entrez Gene ID
654830
Gene Name
leucine-rich repeat LGI family, member 3
Gene Aliases
si:ch211-198o12.2
Chromosome Location
Chr.8: 51375308 - 51404806 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039679.1 NP_001034768.1 1 - 2 310 114
GenBank mRNA AY309263.1 - 1 - 2 310 114
BC162727.1 - 1 - 2 341 114

View Full Product Details