Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dipk1aa
Entrez Gene ID
678606
Gene Name
divergent protein kinase domain 1Aa
Gene Aliases
dipk1a, fam69a, fam69aa, fc67f08, wu:fc67f08, zgc:136922
Chromosome Location
Chr.2: 10642127 - 10687226 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040354.1 NP_001035444.1 2 - 3 439 67
GenBank mRNA BC115333.1 - 2 - 3 439 67

View Full Product Details