Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrrc61
Entrez Gene ID
553439
Gene Name
leucine rich repeat containing 61
Gene Aliases
si:dkey-200j6.2, zgc:136338
Chromosome Location
Chr.25: 25737372 - 25747473 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042754.1 NP_001036219.1 5 - 6 705 116
XM_005163094.4 XP_005163151.1 5 - 6 781 116
GenBank mRNA BC095242.1 - 5 - 6 600 116
BC117587.1 - 5 - 6 705 116

View Full Product Details