Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrc61
Entrez Gene ID
553439
Gene Name
leucine rich repeat containing 61
Gene Aliases
si:dkey-200j6.2, zgc:136338
Chromosome Location
Chr.25: 25737372 - 25747473 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042754.1 NP_001036219.1 7 - 8 828 101
GenBank mRNA BC095242.1 - 7 - 8 723 101
BC117587.1 - 7 - 8 828 101

View Full Product Details