Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnfsf12
Entrez Gene ID
567542
Gene Name
TNF superfamily member 12
Gene Aliases
zgc:153941
Chromosome Location
Chr.7: 268475 - 276794 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076607.1 NP_001070075.1 1 - 2 499 76
GenBank mRNA BC124495.1 - 1 - 2 499 76
BC154255.1 - 1 - 2 187 76
HM439092.1 - 1 - 2 170 76

View Full Product Details