Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
amotl2a
Entrez Gene ID
558920
Gene Name
angiomotin like 2a
Gene Aliases
amotl2, cb308, sb:cb308, wu:fc41b07, wu:fc64c06
Chromosome Location
Chr.6: 27992017 - 27999374 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080177.1 NP_001073646.1 3 - 4 1139 62
GenBank mRNA BC163299.1 - 3 - 4 1013 62
DQ887096.1 - 3 - 4 1139 62

View Full Product Details