Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crtac1a
Entrez Gene ID
559626
Gene Name
cartilage acidic protein 1a
Gene Aliases
ASPIP, cb184, crtac1, sb:cb184, zgc:165343
Chromosome Location
Chr.1: 54933190 - 54947618 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080178.1 NP_001073647.1 6 - 7 871 71
XM_005160175.4 XP_005160232.1 6 - 7 953 71
XM_021475056.1 XP_021330731.1 6 - 7 954 71
XM_021475062.1 XP_021330737.1 6 - 7 922 71
XM_021475068.1 XP_021330743.1 6 - 7 952 71
GenBank mRNA DQ351708.1 - 6 - 7 871 71

View Full Product Details